Learn More
Medchemexpress LLC DPC-681 | 284661-68-3 | 99.72% | 669.85 | 50 MG
SDP
Catalog No. 5000385508
Encompass_Preferred
DPC-681 is a potent and selective inhibitor of HIV protease, effective against wild-type HIV-1. It is intended for research use only.
  • Potent and selective inhibitor of HIV protease.
  • Effective against wild-type HIV-1.
  • Shows no loss in potency toward recombinant mutant HIVs with the D30N mutation.
  • Fivefold or smaller loss in potency toward mutant variants with three to five amino acid substitutions.
  • Suitable for laboratory research applications.
Catalog No. 50-003-85508 Supplier Medchemexpress LLC Supplier No. HY1940050MG
DPC-681 is a potent and selective inhibitor of HIV protease, effective against wild-type HIV-1. It is intended for research use only.
  • Potent and selective inhibitor of HIV protease.
  • Effective against wild-type HIV-1.
  • Shows no loss in potency toward recombinant mutant HIVs with the D30N mutation.
  • Fivefold or smaller loss in potency toward mutant variants with three to five amino acid substitutions.
  • Suitable for laboratory research applications.