Learn More
Medchemexpress LLC DPC-AJ1951 (TFA) | 99.4% | 1666.99 (free base) | 1 MG
SDP
Catalog No. 5000424856
Encompass_Preferred
DPC-AJ1951 TFA is a 14 amino acid peptide that acts as a potent agonist of the parathyroid hormone (PTH)/PTH-related peptide receptor (PPR). Its activity has been characterized in ex vivo and in vivo assays of bone resorption. In vitro, it induces intracellular Ca2+ mobilization in HEK 293 cells transfected with the human PPR.
  • Acts as a potent agonist of the parathyroid hormone (PTH)/PTH-related peptide receptor (PPR)
  • Activity characterized in ex vivo and in vivo assays of bone resorption
  • Induces intracellular Ca2+ mobilization in HEK 293 cells transfected with the human PPR
  • 14 amino acid peptide
Catalog No. 50-004-24856 Supplier Medchemexpress LLC Supplier No. HYP1418A1MG
DPC-AJ1951 TFA is a 14 amino acid peptide that acts as a potent agonist of the parathyroid hormone (PTH)/PTH-related peptide receptor (PPR). Its activity has been characterized in ex vivo and in vivo assays of bone resorption. In vitro, it induces intracellular Ca2+ mobilization in HEK 293 cells transfected with the human PPR.
  • Acts as a potent agonist of the parathyroid hormone (PTH)/PTH-related peptide receptor (PPR)
  • Activity characterized in ex vivo and in vivo assays of bone resorption
  • Induces intracellular Ca2+ mobilization in HEK 293 cells transfected with the human PPR
  • 14 amino acid peptide