Learn More
Perkin Elmer US LLC DPC TFM Cap for High Pressure 100 mL, 100 Bar Digestion Vessels with Pressure Sensor
Catalog No. 502111710
Encompass_Preferred
Includes Vessel Cap made of TFM with pressure sensor for High Pressure 100 mL (100 Bar) Digestion Vessel.
Catalog No. 50-211-1710 Supplier Perkin Elmer US LLC Supplier No. N3133016
Add to Cart
Add to Cart
Request a Quote
Includes Vessel Cap made of TFM with pressure sensor for High Pressure 100 mL (100 Bar) Digestion Vessel.