Learn More
Medchemexpress LLC HY-100937 25mg , DPCPX CAS:102146-07-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10093725MG
Encompass_Preferred
HY-100937 25mg DPCPX CAS: 102146-07-6 DPCPX (PD 116948), a xanthine derivative, is a highly potent and selective Adenosine A1 receptor antagonist, with a Ki of 0.46 nM in 3H-CHA binding to A1 receptors in rat whole brain membranes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1244