Learn More
Aviva Systems Biology Corp MUCL3 Antibody - C-terminal region (ARP50275_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50275_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DPCR1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human DPCR1. Alternate names: MGC126710, MGC126712, PBLT, bCX105N19.6ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60305