Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Tintometer™ DPD 1 Buffer Solution

Catalog No. 09050139 Shop All Tintometer Group Products
Change view
Click to view available options
Quantity:
15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-050-139 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-050-139 Supplier Lovibond™ Supplier No. 471010
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For photometric determination of free/total chlorine in water testing. Lovibond™ Tintometer™ DPD 1 Buffer Solution is an environmentally friendly reagent packaged in a color-coded blue dropper bottle for precise liquid dosing.

For photometric determination of free/total chlorine in water testing. Lovibond™ Tintometer™ DPD 1 Buffer Solution is an environmentally friendly reagent packaged in a color-coded blue dropper bottle for precise liquid dosing.

  • Color coded bottle for quick visual identification of the reagent
  • Convenient dropper bottle provides precise dosing of liquid reagents
  • Environmentally friendly formulation free of boric acid
  • For use with Lovibond™ photometers
  • Carefully developed formula for accurate analysis results

Specifications

Analytes Chlorine
Color Colorless
pH Range 7 at 20°C
Format Aqueous Solution
For Use With (Application) Water sample analysis (e.g. chlorine determination)
For Use With (Equipment) Lovibond™ photometer
Packaging Blue Dropper Bottle
Quantity 15 mL
Range 0.02 to 4mg/L total chlorine
Sample Size 10 mL
Storage Requirements Room temperature unopened, Refrigerated after
Type Buffer solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.