Learn More
Hach Company DPD Free Chlorine Reagent, Swiftest™ Dispenser
Supplier:  Hach Company 2802300
Encompass
DPD Free Chlorine Reagent Dispenser and Reagent Vial. USEPA accepted for reporting drinking water analysis using Hach Method 8021. Total chlorine and potassium iodide Swiftest dispensers also available.Sample Size: 10 mL. Range: 0.02 to 2.00 mg/L Cl2. Contains 1 reagent vial with enough reagent for approximately 250 Tests. *This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Catalog No. 50-199-6697