Learn More
Hach Company DPD Free Chlorine Reagent Powder Pillows, 25 mL pk/1000
Supplier:  Hach Company 1407028
Encompass
For free chlorine determination by the DPD method. USEPA accepted for reporting drinking water analysis using Hach Method 8021. Ranges: LR: up to 2.00 mg/L; MR: 0.05 - 4.50 mg/L; HR: 0 .1- 10.0 mg/L. Sample Sizes: LR: 25 mL; MR: 10 mL; HR: 5 mL. Pack of 1000 powder pillows. Boric Acid free!*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Catalog No. 50-199-4913