Learn More
Hach Company DPD Free Chlorine Reagent Powder Pillows, 25 mL, pk/100
Catalog No. 501994914 Shop All Hach Company Products
Encompass
For free chlorine determination by the DPD method. USEPA accepted for reporting drinking water analysis using Hach Method 8021. Ranges: LR: up to 2.00 mg/L; MR: 0.05-4.00 mg/L; HR: 0.1-10.0 mg/L. Sample Sizes: LR: 25 mL; MR: 10 mL; HR: 5 mL. Pack of 100 powder pillows.Boric Acid free!*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Catalog No. 50-199-4914 Supplier Hach Company Supplier No. 1407099
For free chlorine determination by the DPD method. USEPA accepted for reporting drinking water analysis using Hach Method 8021. Ranges: LR: up to 2.00 mg/L; MR: 0.05-4.00 mg/L; HR: 0.1-10.0 mg/L. Sample Sizes: LR: 25 mL; MR: 10 mL; HR: 5 mL. Pack of 100 powder pillows.Boric Acid free!*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.