Learn More
Hach Company DPD Free Chlorine Reagent Powder Pillows, 5 mL, PK/1000
Supplier:  Hach Company 1407728
Encompass
For determination of free chlorine by the DPD method. For use with Hach's Chlorine Visual Test Kits. Sample Size: 5 mL. Range: up to 3.5 mg/L Cl₂. Pack of 1000 powder pillows. Boric Acid free!*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Catalog No. 50-199-4917