Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ DPD Free Chlorine Reagent Refills
SDPSCI_ED

Catalog No. S72387 Shop All LaMotte Company Products
Change view
Click to view available options
Type:
DPD 1A Liquid Reagent
DPD 1B Liquid Reagent
DPD 3 Liquid Reagent
Volume (English):
1 oz.
2 oz.
Volume (Metric):
30 mL
60 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Volume (English) Volume (Metric)
S72387 DPD 1A Liquid Reagent 1 oz. 30 mL
S72388 DPD 1A Liquid Reagent 2 oz. 60 mL
S72390 DPD 1B Liquid Reagent 2 oz. 60 mL
S72392 DPD 3 Liquid Reagent 2 oz. 60 mL
S72389 DPD 1B Liquid Reagent 1 oz. 30 mL
S72391 DPD 3 Liquid Reagent 1 oz. 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. S72387 Supplier LaMotte™ Supplier No. P6740G
Add to Cart
Add to Cart
This item is not returnable. View return policy

The liquid alternative to DPD tablet refills

  • DPD £1A and DPD £1B test Free Available Chlorine (Cl)
  • DPD £3 is added to the reacted sample to measure Total Cl
  • Comes in 30 mL and 60 mL sizes
  • DPD = N, N-Diethyl-1, 4-phenylene-diamine sulfate

Specifications

Volume (Metric) 30 mL
For Use With (Application) Measurement of total chlorine, provides quicker results and clearer colors than tablets
For Use With (Equipment) LaMotte™ colorimeters and comparators
Quantity Each
Type DPD 1A Liquid Reagent
Volume (English) 1 oz.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.