Learn More
Description
- DPD £1A and DPD £1B test Free Available Chlorine (Cl)
- DPD £3 is added to the reacted sample to measure Total Cl
- Comes in 30 mL and 60 mL sizes
- DPD = N, N-Diethyl-1, 4-phenylene-diamine sulfate
Specifications
Specifications
| Volume (Metric) | 30 mL |
| For Use With (Application) | Measurement of total chlorine, provides quicker results and clearer colors than tablets |
| For Use With (Equipment) | LaMotte™ colorimeters and comparators |
| Quantity | Each |
| Type | DPD 3 Liquid Reagent |
| Volume (English) | 1 oz. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

