Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lovibond™ DPD HR No. 1 and No. 3 HR EVO Combination Pack
Shop All Tintometer Group Products
Click to view available options
No. of Tests:
100 Tests
250 Tests
Description
With over 50 years of manufacturing water quality reagents, Lovibond™ knows choosing the right test for your process or application is critical for accurate results. Each tablet is protected from humidity, heat and other environmental interferences, as pharmaceutical-grade aluminum blister packaging individually seals up to 10 tablets on an individual strip.
- Easy to dispense blister pack makes dispensing tablets safer
- Shelf-stable, analytical solution for testing water quality parameters
- Accurate and precise dosing
- Long shelf-life
Specifications
Specifications
| Analytes | Chlorine |
| Format | Tablet |
| For Use With (Application) | Water Quality Analysis |
| For Use With (Equipment) | Lovibond™ Colorimeters and Spectrophotometers |
| Includes | 100 DPD No. 1 HR and 100 DPD No. 3 HR EVO Tablets |
| No. of Tests | 100 Tests |
| Sample Size | 10 mL |
| Test Method | DPD |
| Test Time | 2 min |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction