Learn More
Hach Company DPD Chlorine Indicator Solution Ampules, ULR, pk/20
Supplier:  Hach Company 2493220
Encompass
For ultra-low Total Chlorine Determination by the DPD method. Hach Method 8370. Range: 2 - 500 μg/L Cl₂. Ampuled for stability. Package of 20x 1.5 mL ampules.Boric Acid free!*This product has not been evaluated to test for chlorine and chlormaines in medical applications in the United States.
Catalog No. 50-199-6073