Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPD Indicator Solution For Residual Chlorine Analysis, Reagents

Catalog No. 01335414
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-335-414 100 mL Glass Bottle
01-335-413 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-335-414 Supplier Reagents Holdings Llc Supplier No. CD280000100C
Add to Cart
Add to Cart

DPD Indicator Solution For Residual Chlorine Analysis, Reagent, for general laboratory use. Recommended Storage: 2°C to 8°C (36°F to 46°F).Manufactured in ISO 9001 facility.

Specifications

Color Colorless to Light Pink
Volume (Metric) 100 mL
Packaging Glass Bottle
Quantity 100 mL
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.