Learn More
Description
- High accuracy
- Blister packaging provides resistance to most climactic conditions and easy, mess-free handling
- Precise reagent dosing
- Significantly longer shelf-life than powders and liquids
Specifications
Specifications
| pH Range | 0 to 14 |
| Volume (Metric) | 10 mL |
| For Use With (Equipment) | Colorimeter |
| No. per Pack | 500 Pk. |
| Type | Free Chlorine Reagent |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.