Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPD No.1 Tablet Reagent, 500 Pk., Lovibond™

Catalog No. 09050156 Shop All Tintometer Group Products
Change view
Click to view available options
Volume (Metric):
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
09-050-156 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-050-156 Supplier Tintometer Group Supplier No. 511052BT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used with Lovibond colorimeters and test kits for the determination of free chlorine, bromine, chlorine dioxide, bromine, and ozone.

  • High accuracy
  • Blister packaging provides resistance to most climactic conditions and easy, mess-free handling
  • Precise reagent dosing
  • Significantly longer shelf-life than powders and liquids

Specifications

pH Range 0 to 14
Volume (Metric) 10 mL
For Use With (Equipment) Colorimeter
No. per Pack 500 Pk.
Type Free Chlorine Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.