Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ DPD No. 3 Evo Tablets

Catalog No. 01671057 Shop All Tintometer Group Products
Change view
Click to view available options
No. of Tests:
100 Tests
250 Tests
500 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Tests
01-671-057 500 Tests
01-671-055 100 Tests
01-671-056 250 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 01-671-057 Supplier Lovibond™ Supplier No. 511422BT
Add to Cart
Add to Cart
This item is not returnable. View return policy

This tablet is used in combination with the DPD No. 1 Tablet to measure Free and Total Chlorine concentrations in a water sample.

  • Shelf-stable, analytical solution for testing water quality parameters with tablet reagents
  • Each tablet is protected from humidity, heat and other environmental interferences
  • Pharmaceutical-grade aluminum blister packaging individually seals up to 10 tablets on an individual strip
  • Easy to dispense blister packaging provides safe handling of the reagents
  • Accurate and precise dosing
  • Long shelf life

Specifications

Analytes Chlorine
Format Tablet
For Use With (Application) Water Quality Analysis
For Use With (Equipment) Lovibond™ Colorimeters and Spectrophotometers
No. of Tests 500 Tests
Sample Size 10 mL
Test Method DPD
Test Time 2 min
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.