Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lovibond™ DPD No. 3 Evo Tablets
Shop All Tintometer Group Products
Click to view available options
No. of Tests:
100 Tests
250 Tests
500 Tests
Description
- Shelf-stable, analytical solution for testing water quality parameters with tablet reagents
- Each tablet is protected from humidity, heat and other environmental interferences
- Pharmaceutical-grade aluminum blister packaging individually seals up to 10 tablets on an individual strip
- Easy to dispense blister packaging provides safe handling of the reagents
- Accurate and precise dosing
- Long shelf life
Specifications
Specifications
| Analytes | Chlorine |
| Format | Tablet |
| For Use With (Application) | Water Quality Analysis |
| For Use With (Equipment) | Lovibond™ Colorimeters and Spectrophotometers |
| No. of Tests | 100 Tests |
| Sample Size | 10 mL |
| Test Method | DPD |
| Test Time | 2 min |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction