Learn More
HF Scientific Inc Total Chlorine DPD Reagent Dispenser, 400 Tests (2x200)
Catalog No. NC1736374
Encompass_Preferred
This HF scientific portable dispenser accurately delivers the correct volume of DPD reagent for total chlorine. It is supplied with enough DPD powder for 400 tests. To use, simply invert the dispenser over the sample and press the button to dispense the reagent. The powder dissolves quickly in the sample, leaving no messy residue behind. It is a 10ml sample size, with formulations for use with HF scientific and Hach photometers.
Catalog No. NC1736374 Supplier HF Scientific Inc Supplier No. 10448A
This HF scientific portable dispenser accurately delivers the correct volume of DPD reagent for total chlorine. It is supplied with enough DPD powder for 400 tests. To use, simply invert the dispenser over the sample and press the button to dispense the reagent. The powder dissolves quickly in the sample, leaving no messy residue behind. It is a 10ml sample size, with formulations for use with HF scientific and Hach photometers.