Learn More
Specifications
Specifications
| Color | Clear, colorless to slight yellow |
| Format | Liquid |
| Packaging | Amber Glass |
| Quantity | 1 L |
| Storage Requirements | Refrigerate |
| Type | DPD Solution |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.