Learn More
AdipoGen DPD sulfate
SDP
Catalog No. 502561022
Encompass_Preferred
Chemical. CAS 6283-63-2. Formula C10H18N2O4S. MW 262.33. N,N-Diethyl-p-phenylenediamine sulfate DPD is used as a chromogenic indicator or reagent in analytical chemistry and in water quality testing. It is commonly used in a test called the DPD method, primarily for the measurement of free and total chlorine by titrimetric, colorimetric and spectroscopic methods in water. DPD reacts with free chlorine hypochlorous acid and hypochlorite ions to form a pink or magenta-colored compound. The intensity of the color is directly proportional to the concentration of chlorine, making it a convenient method for chlorine quantification. In general, it is used for spectrophotometric determination KIO4 levels, drugs containing phenolic groups or aromatic amino groups and as a peroxidase substrate. DPD has been developed as a color developing agent in photgraphy.
Catalog No. 50-256-1022 Supplier AdipoGen Supplier No. CDXD0932G250
Chemical. CAS 6283-63-2. Formula C10H18N2O4S. MW 262.33. N,N-Diethyl-p-phenylenediamine sulfate DPD is used as a chromogenic indicator or reagent in analytical chemistry and in water quality testing. It is commonly used in a test called the DPD method, primarily for the measurement of free and total chlorine by titrimetric, colorimetric and spectroscopic methods in water. DPD reacts with free chlorine hypochlorous acid and hypochlorite ions to form a pink or magenta-colored compound. The intensity of the color is directly proportional to the concentration of chlorine, making it a convenient method for chlorine quantification. In general, it is used for spectrophotometric determination KIO4 levels, drugs containing phenolic groups or aromatic amino groups and as a peroxidase substrate. DPD has been developed as a color developing agent in photgraphy.