Learn More
Hach Company DPD Total Chlorine Reagent Accuvac Ampules, pk/25
Supplier:  Hach Company 2503025
Encompass
For determination of Total Chlorine Reagent by the DPD method, using Accuvac Ampules. USEPA accepted for reporting drinking and wastewater analysis (Hach Method 8167). Range: 0.02 - 2.00 mg/L Cl2. Package of 25 ampules.*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Catalog No. 50-199-6100