Learn More
Hach Company DPD Total Chlorine Reagent Powder Pillows, 25 mL, pk/100
Catalog No. 501994910 Shop All Hach Company Products
Encompass
For total chlorine determination by the DPD method. USEPA accepted for reporting drinking and wastewater analysis using Hach Method 8167. Ranges: Low Range: up to - 2.00 mg/L; Mid Range: 0.05-4.00 mg/L; High Range: 0.1-10.0 mg/L. Sample Size: Low Range - 25 mL; Mid Range: 10 mL; High Range: 5 mL. Pack of 100 powder pillows.Boric Acid free! *This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
This item is not returnable. View return policy
Catalog No. 50-199-4910 Supplier Hach Company Supplier No. 1406499
Add to Cart
Add to Cart
For total chlorine determination by the DPD method. USEPA accepted for reporting drinking and wastewater analysis using Hach Method 8167. Ranges: Low Range: up to - 2.00 mg/L; Mid Range: 0.05-4.00 mg/L; High Range: 0.1-10.0 mg/L. Sample Size: Low Range - 25 mL; Mid Range: 10 mL; High Range: 5 mL. Pack of 100 powder pillows.Boric Acid free! *This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.