Learn More
Hach Company DPD Total Chlorine Reagent Powder Pillows, 5 mL, pk/100
Supplier:  Hach Company 1407699
Encompass
For determination of total chlorine by the DPD method. For use with Hach's Chlorine Visual Test Kits. Sample Size: 5 mL. Range: up to 3.5 mg/L Cl₂. Pack of 100 powder pillows.Boric Acid free! *This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Catalog No. 50-199-4916