Learn More
Hach Company DPD Total Chlorine Reagent Powder Pillows, 5 mL, pk/1000
Catalog No. 501994915 Shop All Hach Company Products
Encompass
For determination of total chlorine by the DPD method. For use with Hach's Chlorine Visual Test Kits. Sample Size: 5 mL. Range: up to 3.5 mg/L Cl₂. Pack of 1000 powder pillows. Boric Acid free!*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
This item is not returnable. View return policy
Catalog No. 50-199-4915 Supplier Hach Company Supplier No. 1407628
For determination of total chlorine by the DPD method. For use with Hach's Chlorine Visual Test Kits. Sample Size: 5 mL. Range: up to 3.5 mg/L Cl₂. Pack of 1000 powder pillows. Boric Acid free!*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.