Learn More
Hach Company DPD Total Chlorine Reagent, Swiftest™ Dispenser
Supplier:  Hach Company 2802400
Encompass
DPD Total Chlorine Reagent Dispenser and reagent vial, approximately 250 Tests. USEPA accepted for reporting drinking water and wastewater analyses using Hach Method 8167. Free chlorine and potassium iodide Swiftest dispensers also available.Sample Size: 10 mL. Range: 0.02 to 2.00 mg/L. Contains 1 refill vial with enough reagent for approximately 250 tests.*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Catalog No. 50-199-6698