Learn More
Aqua Solutions DPD Total Chlorine Reagent Powder Pillows, 25 mL, pk/1000
SDP
Catalog No. NC1448666
Encompass
For total chlorine determination by the DPD method. USEPA accepted for reporting drinking and wastewater analysis using Hach Method 8167. Ranges: Low Range: up to - 2.00 mg/L; Mid Range: 0.05 - 4.00 mg/L; High Range: 0 .1- 10.0 mg/L. Sample Size: Low Range - 25 mL. Mid Range: 10 mL. High Range: 5 mL. Pack of 1000 powder pillows. Boric Acid free!*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Catalog No. NC1448666 Supplier Aqua Solutions Supplier No. HA1406428
For total chlorine determination by the DPD method. USEPA accepted for reporting drinking and wastewater analysis using Hach Method 8167. Ranges: Low Range: up to - 2.00 mg/L; Mid Range: 0.05 - 4.00 mg/L; High Range: 0 .1- 10.0 mg/L. Sample Size: Low Range - 25 mL. Mid Range: 10 mL. High Range: 5 mL. Pack of 1000 powder pillows. Boric Acid free!*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.