Learn More
Hach Company DPD Total Chlorine Reagent Powder Pillows, 5 mL, pk/50
Supplier:  Hach Company 2198246
Encompass
For determination of total chlorine with Hach's Chlorine Visual Test Kits- 2060400. Sample Size: 5 mL. Range: up to 2.5 mg/L Cl₂. Pack of 50 Powder Pillows.Boric Acid free!*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Catalog No. 50-199-5548