Learn More
Hach Company DPD Total Chlorine Swiftest™ Dispenser Refill Vial, approximately 250 Tests
Catalog No. 501995396 Shop All Hach Company Products
Encompass
For total chlorine determination by the DPD Total Chlorine method. For use with Hach Swiftest DPD powder dispenser. USEPA accepted for reporting for drinking and wastewater analysis using Hach Method 8167. Free chlorine and potassium iodide Swiftest refill vials also available.Range: 0.02 - 2.00 mg/L Cl2. Sample size: 10 mL. Approximately 250 tests per vial. *This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Catalog No. 50-199-5396 Supplier Hach Company Supplier No. 2105660
For total chlorine determination by the DPD Total Chlorine method. For use with Hach Swiftest DPD powder dispenser. USEPA accepted for reporting for drinking and wastewater analysis using Hach Method 8167. Free chlorine and potassium iodide Swiftest refill vials also available.Range: 0.02 - 2.00 mg/L Cl2. Sample size: 10 mL. Approximately 250 tests per vial. *This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.