Learn More
CPC Scientific H-Tyr-DPen-Gly-Phe-DPen-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific ENKP001B
Encompass
Peptide that performs direct antinociceptive actions via the delta1-opioid receptor, while modulatory via the delta2-opioid receptor.SEQUENCE: H-Tyr-DPen-Gly-Phe-DPen-OH (trifluoroacetate salt)(DPen2 and DPen5 bridge)ONE-LETTER SEQUENCE: Y-DPen-GF-DPen(DPen2&DPen5 bridge)MOLECULAR FORMULA: C30H39N5O7S2MOLECULAR WEIGHT: 645.8STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [88373-73-3]SYNONYMS: RESEARCH AREA: painREFERENCES:H.I. Mosberg et al., PNAS, 80, 5871 (1983)SKU(s): ENKP-001A, ENKP-001B
Catalog No. 50-210-9249