Learn More
Abcam DPDPE, delta opioid receptor agonist, 5MG
Catalog No. 502707359
Encompass_Preferred
MW 645.79 g/mol, Purity >95%. Potent and selective δ opioid receptor agonist (Ki values are 14, >1000 and >1000 nM for opioid receptor subtypes δ, κ and μ, respectively). Centrally active following systemic administration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7359 Supplier Abcam Supplier No. AB1206755MG
MW 645.79 g/mol, Purity >95%. Potent and selective δ opioid receptor agonist (Ki values are 14, >1000 and >1000 nM for opioid receptor subtypes δ, κ and μ, respectively). Centrally active following systemic administration.
The product is subject to the following: Abcam Restricted Use Statement