Learn More
Cayman Chemical dPE-MPEG 5000 250mg
SDP
Catalog No. 502950869
Encompass_Preferred
A PEGylated form of 1,2-DSPE
Catalog No. 50-295-0869 Supplier Cayman Chemical Supplier No. 34652250 MG
A PEGylated form of 1,2-DSPE