Learn More
Cayman Chemical dPE-PEG 2000-CoH 25mg
SDP
Catalog No. 502945811
Encompass_Preferred
A PEGylated derivative of DSPE; has been used to link transferrin receptors to the surface of liposomes encapsulating doxorubicin for targeted delivery of doxorubicin in vitro and in vivo; nanomicelles containing DSPE-PEG(2000)-COOH linked to borneol and encapsulating doxorubicin reduce tumor growth and metastasis in an in situ C6 murine model of glioblastoma
Catalog No. 50-294-5811 Supplier Cayman Chemical Supplier No. 4216425 MG
A PEGylated derivative of DSPE; has been used to link transferrin receptors to the surface of liposomes encapsulating doxorubicin for targeted delivery of doxorubicin in vitro and in vivo; nanomicelles containing DSPE-PEG(2000)-COOH linked to borneol and encapsulating doxorubicin reduce tumor growth and metastasis in an in situ C6 murine model of glioblastoma