Learn More
Cayman Chemical DSPE-PEG(2000)-COOH, Purity: ≥95%, 5 mg
SDP
Catalog No. 502944311
Encompass_Preferred
A PEGylated derivative of DSPE; has been used to link transferrin receptors to the surface of liposomes encapsulating doxorubicin for targeted delivery of doxorubicin in vitro and in vivo; nanomicelles containing DSPE-PEG(2000)-COOH linked to borneol and encapsulating doxorubicin reduce tumor growth and metastasis in an in situ C6 murine model of glioblastoma
Catalog No. 50-294-4311 Supplier Cayman Chemical Supplier No. 421645 MG
A PEGylated derivative of DSPE; has been used to link transferrin receptors to the surface of liposomes encapsulating doxorubicin for targeted delivery of doxorubicin in vitro and in vivo; nanomicelles containing DSPE-PEG(2000)-COOH linked to borneol and encapsulating doxorubicin reduce tumor growth and metastasis in an in situ C6 murine model of glioblastoma