Learn More
Cayman Chemical dPE-PEG 2000 Cynur 500ug
SDP
Catalog No. 502915699
Encompass_Preferred
A PEGylated form of DSPE; has been used in the generation of LNPs for the delivery of miRNA in vitro; LNPs containing DSPE-PEG(2000) cyanur, encapsulating let-7a miRNA, and conjugated to ephrin-A1 reduce the proliferation and colony formation and size of NCI H2081, NCI 5380, and A549 cancer cells, as well as reduce migration of NCI 5380 cells in a wound healing assay
Catalog No. 50-291-5699 Supplier Cayman Chemical Supplier No. 37797500 UG
A PEGylated form of DSPE; has been used in the generation of LNPs for the delivery of miRNA in vitro; LNPs containing DSPE-PEG(2000) cyanur, encapsulating let-7a miRNA, and conjugated to ephrin-A1 reduce the proliferation and colony formation and size of NCI H2081, NCI 5380, and A549 cancer cells, as well as reduce migration of NCI 5380 cells in a wound healing assay