Learn More
Cayman Chemical dPE-PEG 2000 MaleImIde 50mg
SDP
Catalog No. 502945689
Encompass_Preferred
A MEK inhibitor (IC50 5.73 nM) and an active metabolite of CI-1040 reduces viral titers in A549 and MDCK cells infected with influenza A (EC50s 6.356 and 4.498 UM respectively) increases survival in H1N1pdm09 influenza A-infected mice at 50 mg/kg per day
Catalog No. 50-294-5689 Supplier Cayman Chemical Supplier No. 4201350 MG
A MEK inhibitor (IC50 5.73 nM) and an active metabolite of CI-1040 reduces viral titers in A549 and MDCK cells infected with influenza A (EC50s 6.356 and 4.498 UM respectively) increases survival in H1N1pdm09 influenza A-infected mice at 50 mg/kg per day