Learn More
Cayman Chemical dPE-PEG 2000 MaleImIde 5mg
SDP
Catalog No. 502945632
Encompass_Preferred
A metabolite of linoleic acid increases transactivation of PPARa and PPARY in reporter assays using mouse aortic endothelial cells at 68 UM induces chemotaxis of primary human monocytes at 10 UM has been found in oxLDL
Catalog No. 50-294-5632 Supplier Cayman Chemical Supplier No. 420135 MG
A metabolite of linoleic acid increases transactivation of PPARa and PPARY in reporter assays using mouse aortic endothelial cells at 68 UM induces chemotaxis of primary human monocytes at 10 UM has been found in oxLDL