Learn More
Cayman Chemical dPE-PEG 2000-NHS 50mg
SDP
Catalog No. 502917693
Encompass_Preferred
A PEGylated derivative of DSPE; has been used in the generation of micelles and in combination with other lipids in the formation of liposomes; intravenous injection of liposomes containing DSPE-PEG(2000)-NHS ester localize to mouse liver and tumor tissue; liposomes containing DSPE-PEG(2000)-NHS ester conjugated to H6 peptide and encapsulating doxorubicin reduce tumor growth in an SK-BR-3 breast cancer mouse xenograft model
Catalog No. 50-291-7693 Supplier Cayman Chemical Supplier No. 3727550 MG
A PEGylated derivative of DSPE; has been used in the generation of micelles and in combination with other lipids in the formation of liposomes; intravenous injection of liposomes containing DSPE-PEG(2000)-NHS ester localize to mouse liver and tumor tissue; liposomes containing DSPE-PEG(2000)-NHS ester conjugated to H6 peptide and encapsulating doxorubicin reduce tumor growth in an SK-BR-3 breast cancer mouse xenograft model