Learn More
Cayman Chemical dPE-PolysarcosIn150 10mg
SDP
Catalog No. 502918641
Encompass_Preferred
An amine-functionalized hydrophilic polymer; a polysarcosine-labeled form of DSPE; polysarcosines have been used as alternatives to PEGylation in LNPs and to reduce the immunogenicity of administered proteins in vivo
Catalog No. 50-291-8641 Supplier Cayman Chemical Supplier No. 3904510 MG
An amine-functionalized hydrophilic polymer; a polysarcosine-labeled form of DSPE; polysarcosines have been used as alternatives to PEGylation in LNPs and to reduce the immunogenicity of administered proteins in vivo