Learn More
Cayman Chemical dPE-PolysarcosIn50 10mg
SDP
Catalog No. 502918596
Encompass_Preferred
An inhibitor of mitochondrial dysfunction increases intracellular ATP levels in Hep3B cells at a 3 UM rescues LPS-induced apoptosis and decreases in the mitochondrial membrane potential in BV-2 microglial cells at 5 UM protects against BSO-induced decreases in cell viability in fibroblasts isolated from patients with various mitochondrial diseases (EC50s 2.3-5.1 UM) in an electron transport chain-independent manner reduces plasma levels of creatinine and decreases acute renal tubular necrosis in a mouse model of bilateral retroperitoneal renal ischemia-reperfusion injury at 50 mg/kg
Catalog No. 50-291-8596 Supplier Cayman Chemical Supplier No. 3904310 MG
An inhibitor of mitochondrial dysfunction increases intracellular ATP levels in Hep3B cells at a 3 UM rescues LPS-induced apoptosis and decreases in the mitochondrial membrane potential in BV-2 microglial cells at 5 UM protects against BSO-induced decreases in cell viability in fibroblasts isolated from patients with various mitochondrial diseases (EC50s 2.3-5.1 UM) in an electron transport chain-independent manner reduces plasma levels of creatinine and decreases acute renal tubular necrosis in a mouse model of bilateral retroperitoneal renal ischemia-reperfusion injury at 50 mg/kg