Learn More
Strem, An Ascensus Company CAS# 166330-10-5. 100G. Bis(2-diphenylphosphinophenyl)ether, 98% DPEphos. MFCD00233863
SDP
Catalog No. 501963538
Encompass
CAS# 166330-10-5. 100G. Bis(2-diphenylphosphinophenyl)ether, 98% DPEphos. MFCD00233863. Molecular Formula: (C6H5)2PC6H4OC6H4P(C6H5)2. Molecular Weight: 538.56. Color/Form: off-white pwdr. Strem# 15-0380. www.strem.com/catalog/v3/15-0380/
Catalog No. 50-196-3538 Supplier Strem, An Ascensus Company Supplier No. 150380100G
CAS# 166330-10-5. 100G. Bis(2-diphenylphosphinophenyl)ether, 98% DPEphos. MFCD00233863. Molecular Formula: (C6H5)2PC6H4OC6H4P(C6H5)2. Molecular Weight: 538.56. Color/Form: off-white pwdr. Strem# 15-0380. www.strem.com/catalog/v3/15-0380/