Learn More
Aviva Systems Biology Corp DPF1 Antibody - middle region (ARP34397_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34397_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DPF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DPF1. Alternate names: NEUD4, BAF45b, neuro-d4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-52953