Learn More
Aviva Systems Biology Corp DPF2 Antibody - N-terminal region (ARP38768_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38768_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against DPF2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DPF2. Alternate names: REQ, UBID4, ubi-d4ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56789