Learn More
Aviva Systems Biology Corp DPF2 Antibody - middle region (ARP38769_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38769_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DPF2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DPF2. Alternate names: MGC10180, REQ, UBID4, ubi-d4ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56790