Learn More
Aviva Systems Biology Corp Dpf3 Antibody - N-terminal region (ARP38943_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38943_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Dpf3. It was validated on Western Blot. . Alternate names: 2810403B03Rik, C78788, CERD4, cer-d4, BAF45C, 6530402L11RikApplicationWB, IHC. Species reactivity: Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-767