Learn More
Medchemexpress LLC HY-12070 10mg Medchemexpress, DPH CAS:484049-04-9 Purity:>98%
SDP
Catalog No. 502080951
Encompass_Preferred
Medchemexpress, HY-12070 10mg DPH CAS:484049-04-9 DPH is a potent cell permeable c-Abl activator, which displays potent enzymatic and cellular activity in stimulating c-Abl activation.   Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0951 Supplier Medchemexpress LLC Supplier No. HY1207010MG
Medchemexpress, HY-12070 10mg DPH CAS:484049-04-9 DPH is a potent cell permeable c-Abl activator, which displays potent enzymatic and cellular activity in stimulating c-Abl activation.   Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.