Learn More
TARGETMOL CHEMICALS INC DPH 5MG
SDP
Catalog No. 502581247
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DPH is a potent cell permeable activator of c-Abl. It shows potent enzymatic and cellular activity in stimulating c-Abl activation. purity: 99%
Catalog No. 50-258-1247 Supplier TARGETMOL CHEMICALS INC Supplier No. T151645MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DPH is a potent cell permeable activator of c-Abl. It shows potent enzymatic and cellular activity in stimulating c-Abl activation. purity: 99%