Learn More
Aviva Systems Biology Corp DPH1 Antibody - middle region (ARP51956_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51956_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DPH1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DPH1. Alternate names: DPH2L, DPH2L1, FLJ33211, OVCA1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-60653