Learn More
Aviva Systems Biology Corp DPH4 Antibody - N-terminal region (ARP53399_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53399_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DPH4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DPH4. Alternate names: JJJ3, ZCSL3, DPH4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61016